N-Methoxy-N-methylcycloheptanecarboxamide

Lieferant: Apollo Scientific

OR451138-1G
APOSOR451138-1GEA 771 EUR
APOSOR451138-1G
N-Methoxy-N-methylcycloheptanecarboxamide
N-Methoxy-N-methylcycloheptanecarboxamide
Formel: C₁₀H₁₉NO₂
Molecular Weight: 185.263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12546238
CAS-Nummer: 253429-08-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR