2,4-Dichloro-6-methyl-quinoline

Lieferant: Apollo Scientific

Synonyme: 2,4-Dichloro-6-methylquinoline

OR451140-1G
APOSOR451140-1GEA 109 EUR
APOSOR451140-1G
2,4-Dichloro-6-methyl-quinoline
2,4-Dichloro-6-methyl-quinoline
Formel: C₁₀H₇Cl₂N
Molecular Weight: 212,075 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02599423
CAS-Nummer: 102878-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR