2,4-Dichloro-6-methoxy-quinoline

Lieferant: Apollo Scientific

Synonyme: 2,4-Dichloro-6-methoxyquinoline

OR451141-1G
APOSOR451141-1GEA 960 EUR
APOSOR451141-1G
2,4-Dichloro-6-methoxy-quinoline
2,4-Dichloro-6-methoxy-quinoline
Formel: C₁₀H₇Cl₂NO
Molecular Weight: 228,075 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03285967
CAS-Nummer: 70049-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR