11-chloro-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline

Lieferant: Apollo Scientific

OR451143-1G
APOSOR451143-1GEA 1740 EUR
APOSOR451143-1G
11-chloro-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline
11-chloro-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline
Formel: C₁₄H₁₄ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD11849287
CAS-Nummer: 5778-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR