(+/-)-1,2,3,4-Tetrahydroisoquinoline-3-methanol

Lieferant: Apollo Scientific

OR451144-1G
APOSOR451144-1GEA 0 EUR
APOSOR451144-1G
(+/-)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
(+/-)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Formel: C₁₀H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD02259418
CAS-Nummer: 63006-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR