(R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol

Lieferant: Apollo Scientific

Synonyme: (R)-(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol

OR451146-1G OR451146-25G OR451146-5G
APOSOR451146-1GEA 147 EUR
APOSOR451146-1G APOSOR451146-25G APOSOR451146-5G
(R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
(R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Formel: C₁₀H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD02684327
CAS-Nummer: 62855-02-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR