4-Spiroindanepiperidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 2,3-Dihydrospiro[indene-1,4'-piperidine] hydrochloride

OR451148-1G
APOSOR451148-1GEA 102 EUR
APOSOR451148-1G
4-Spiroindanepiperidine hydrochloride
4-Spiroindanepiperidine hydrochloride
Formel: C₁₃H₁₈ClN
Molecular Weight: 223,75 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08461061
CAS-Nummer: 96651-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR