8-(Iodomethyl)-1,4-dioxaspiro[4.5]decane

Lieferant: Apollo Scientific

OR451151-1G
APOSOR451151-1GEA 960 EUR
APOSOR451151-1G
8-(Iodomethyl)-1,4-dioxaspiro[4.5]decane
8-(Iodomethyl)-1,4-dioxaspiro[4.5]decane
Formel: C₉H₁₅IO₂
Molecular Weight: 282,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26387804
CAS-Nummer: 625114-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR