1,4-Dioxaspiro[4.5]decane-8-carboxaldehyde

Lieferant: Apollo Scientific

OR451152-1G
APOSOR451152-1GEA 960 EUR
APOSOR451152-1G
1,4-Dioxaspiro[4.5]decane-8-carboxaldehyde
1,4-Dioxaspiro[4.5]decane-8-carboxaldehyde
Formel: C₉H₁₄O₃
Molecular Weight: 170,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847774
CAS-Nummer: 93245-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR