1,4-Dioxaspiro[4.5]decane-8-acetic acid

Lieferant: Apollo Scientific

OR451153-5G
APOSOR451153-5GEA 2850 EUR
APOSOR451153-5G
1,4-Dioxaspiro[4.5]decane-8-acetic acid
1,4-Dioxaspiro[4.5]decane-8-acetic acid
Formel: C₁₀H₁₆O₄
Molecular Weight: 200,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16620931
CAS-Nummer: 134136-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR