8-Methylene-1,4-dioxa-spiro[4.5]decane

Lieferant: Apollo Scientific

Synonyme: 8-Methylene-1,4-dioxaspiro[4.5]decane

OR451154-1G
APOSOR451154-1GEA 134 EUR
APOSOR451154-1G
8-Methylene-1,4-dioxa-spiro[4.5]decane
8-Methylene-1,4-dioxa-spiro[4.5]decane
Formel: C₉H₁₄O₂
Molecular Weight: 154,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779440
CAS-Nummer: 51656-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR