1,3-Diazaspiro[4.5]decane-2,4,8-trione

Lieferant: Apollo Scientific

OR451155-1G
APOSOR451155-1GEA 873 EUR
APOSOR451155-1G
1,3-Diazaspiro[4.5]decane-2,4,8-trione
1,3-Diazaspiro[4.5]decane-2,4,8-trione
Formel: C₈H₁₀N₂O₃
Molecular Weight: 182,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20729251
CAS-Nummer: 51145-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR