1-(1,4-Dioxaspiro[4.5]dec-8-yl)-ethanone

Lieferant: Apollo Scientific

Synonyme: 1-(1,4-Dioxaspiro[4.5]decan-8-yl)ethanone

OR451156-1G
APOSOR451156-1GEA 301 EUR
APOSOR451156-1G
1-(1,4-Dioxaspiro[4.5]dec-8-yl)-ethanone
1-(1,4-Dioxaspiro[4.5]dec-8-yl)-ethanone
Formel: C₁₀H₁₆O₃
Molecular Weight: 184,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20257858
CAS-Nummer: 35477-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR