3-(Cyclopropylmethoxy)-benzoic acid

Lieferant: Apollo Scientific

OR451161-1G
APOSOR451161-1GEA 915 EUR
APOSOR451161-1G
3-(Cyclopropylmethoxy)-benzoic acid
3-(Cyclopropylmethoxy)-benzoic acid
Formel: C₁₁H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12138595
CAS-Nummer: 1047680-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR