2-(Cyclopropylmethoxy)-benzoic acid

Lieferant: Apollo Scientific

OR451162-1G
APOSOR451162-1GEA 579 EUR
APOSOR451162-1G
2-(Cyclopropylmethoxy)-benzoic acid
2-(Cyclopropylmethoxy)-benzoic acid
Formel: C₁₁H₁₂O₃
Molecular Weight: 192.211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06208775
CAS-Nummer: 136013-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR