a-Cyclopropyl-4-chloro-benzylamine
Lieferant: Apollo Scientific
Synonyme:
1-(4-Chlorophenyl)-1-cyclopropylmethylamine
OR451163-250MG
APOSOR451163-250MGEA
579
EUR
APOSOR451163-250MG
a-Cyclopropyl-4-chloro-benzylamine
a-Cyclopropyl-4-chloro-benzylamine
Formel:
C₁₀H₁₂ClN Molecular Weight: 181,662 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05191965 CAS-Nummer: 123312-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...