Boc-3-Cyclopropylalanine
Lieferant: Apollo Scientific
|
Achtung
|
OR451164-5G
APOSOR451164-5GEA
470
EUR
APOSOR451164-5G
Boc-3-Cyclopropylalanine
Boc-3-Cyclopropylalanine
Formel:
C₁₁H₁₉NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061239 CAS-Nummer: 888323-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...