Boc-2-Cyclopropylglycine

Lieferant: Apollo Scientific

Synonyme: 2-((tert-Butoxycarbonyl)amino)-2-cyclopropylacetic acid, Boc-DL-Cyclopropyl-Gly-OH

OR451165-5G
APOSOR451165-5GEA 556 EUR
APOSOR451165-5G
Boc-2-Cyclopropylglycine
Boc-2-Cyclopropylglycine
Formel: C₁₀H₁₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01861823
CAS-Nummer: 54256-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR