3-Cyclopropylalanine

Lieferant: Apollo Scientific

Synonyme: 2-Amino-3-cyclopropylpropanoic acid

OR451166-5G
APOSOR451166-5GEA 346 EUR
APOSOR451166-5G
3-Cyclopropylalanine
3-Cyclopropylalanine
Formel: C₆H₁₁NO₂
Molecular Weight: 129,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02666085
CAS-Nummer: 15785-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR