2-Cyclopropylglycine

Lieferant: Apollo Scientific

Synonyme: 2-Amino-2-cyclopropylacetic acid

OR451167-5G
APOSOR451167-5GEA 771 EUR
APOSOR451167-5G
2-Cyclopropylglycine
2-Cyclopropylglycine
Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01861822
CAS-Nummer: 15785-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR