Cyclopropyl(3-methoxybenzyl)amine

Lieferant: Apollo Scientific

OR451168-1G
APOSOR451168-1GEA 487 EUR
APOSOR451168-1G
Cyclopropyl(3-methoxybenzyl)amine
Cyclopropyl(3-methoxybenzyl)amine
Formel: C₁₁H₁₅NO
Molecular Weight: 177,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07409182
CAS-Nummer: 625437-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR