1-(Cyclobutylcarbonyl)piperazine hydrochloride

Lieferant: Apollo Scientific

OR451169-1G
APOSOR451169-1GEA 510 EUR
APOSOR451169-1G
1-(Cyclobutylcarbonyl)piperazine hydrochloride
1-(Cyclobutylcarbonyl)piperazine hydrochloride
Formel: C₉H₁₆N₂O·ClH
Molecular Weight: 204,697 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975205
CAS-Nummer: 1428443-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR