2-Cyano-N-cyclobutyl-acetamide

Lieferant: Apollo Scientific

OR451170-1G
APOSOR451170-1GEA 684 EUR
APOSOR451170-1G
2-Cyano-N-cyclobutyl-acetamide
2-Cyano-N-cyclobutyl-acetamide
Formel: C₇H₁₀N₂O
Molecular Weight: 138,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16755845
CAS-Nummer: 1064448-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR