1-Phenylcyclobutanamine

Lieferant: Apollo Scientific

OR451172-100MG OR451172-1G OR451172-250MG
APOSOR451172-100MGEA 256 EUR
APOSOR451172-100MG APOSOR451172-1G APOSOR451172-250MG
1-Phenylcyclobutanamine
1-Phenylcyclobutanamine
Formel: C₁₀H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD08444192
CAS-Nummer: 17380-77-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR