4-(1H-Pyrazol-1-yl)-piperidine dihydrochloride
Lieferant: Apollo Scientific
OR451177-5G
APOSOR451177-5GEA
1740
EUR
APOSOR451177-5G
4-(1H-Pyrazol-1-yl)-piperidine dihydrochloride
4-(1H-Pyrazol-1-yl)-piperidine dihydrochloride
Formel:
C₈H₁₃N₃·2ClH Molecular Weight: 224,131 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11858004 CAS-Nummer: 1181458-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...