Methyl-1-benzyl-5-oxopyrrolidine-3-carboxylate
Lieferant: Apollo Scientific
Synonyme:
Methyl 1-Benzyl-5-oxo-3-pyrrolidinecarboxylate
OR451179-100G
APOSOR451179-100GEA
139
EUR
APOSOR451179-100G
Methyl-1-benzyl-5-oxopyrrolidine-3-carboxylate
Methyl-1-benzyl-5-oxopyrrolidine-3-carboxylate
Formel:
C₁₃H₁₅NO₃ Molecular Weight: 233,267 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003196 CAS-Nummer: 51535-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...