Boc-Pro-N(OMe)Me

Lieferant: Apollo Scientific

Synonyme: (S)-1-Boc-2-[methoxy(methyl)carbamoyl]pyrrolidine

OR451181-1G
APOSOR451181-1GEA 388 EUR
APOSOR451181-1G
Boc-Pro-N(OMe)Me
Boc-Pro-N(OMe)Me
Formel: C₁₂H₂₂N₂O₄
Molecular Weight: 258,3 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 115186-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR