(S)-3-Benzyloxypyrrolidine hydrochloride

Lieferant: Apollo Scientific

OR451183-5G
APOSOR451183-5GEA 800 EUR
APOSOR451183-5G
(S)-3-Benzyloxypyrrolidine hydrochloride
(S)-3-Benzyloxypyrrolidine hydrochloride
Formel: C₁₁H₁₅NO·ClH
Molecular Weight: 213.704 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784038
CAS-Nummer: 931409-74-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR