3-Amino-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Amino-1-Boc-pyrrolidine-3-carboxylic acid, 3-amino-1-[(tert-butoxy)carbonyl]pyrrolidine-3-carboxylic acid

OR451184-1G
APOSOR451184-1GEA 95.7 EUR
APOSOR451184-1G
3-Amino-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid
3-Amino-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid
Formel: C₁₀H₁₈N₂O₄
Molecular Weight: 230,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115499
CAS-Nummer: 862372-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR