(S)-N-Methyl-2-pyrrolidinecarboxamide hydrochloride

Lieferant: Apollo Scientific

Synonyme: (S)-N-Methylpyrrolidine-2-carboxamide hydrochloride

OR451186-5G
APOSOR451186-5GEA 289 EUR
APOSOR451186-5G
(S)-N-Methyl-2-pyrrolidinecarboxamide hydrochloride
(S)-N-Methyl-2-pyrrolidinecarboxamide hydrochloride
Formel: C₆H₁₂N₂O·ClH
Molecular Weight: 164,633 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153461
CAS-Nummer: 33208-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR