(S)-N,N-Dimethyl-2-pyrrolidinecarboxamide hydrochloride

Lieferant: Apollo Scientific

OR451187-5G
APOSOR451187-5GEA 411 EUR
APOSOR451187-5G
(S)-N,N-Dimethyl-2-pyrrolidinecarboxamide hydrochloride
(S)-N,N-Dimethyl-2-pyrrolidinecarboxamide hydrochloride
Formel: C₇H₁₄N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD17215582
CAS-Nummer: 149596-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR