1-[(2S)-2-Pyrrolidinylmethyl]-piperidine dihydrochloride
Lieferant: Apollo Scientific
OR451188-1G
APOSOR451188-1GEA
1110
EUR
APOSOR451188-1G
1-[(2S)-2-Pyrrolidinylmethyl]-piperidine dihydrochloride
1-[(2S)-2-Pyrrolidinylmethyl]-piperidine dihydrochloride
Formel:
C₁₀H₂₀N₂·2ClH Molecular Weight: 241,201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11974893 CAS-Nummer: 215918-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...