(R)-1-[(Pyrrolidin-2-yl)methyl]piperidine dihydrochloride

Lieferant: Apollo Scientific

OR451190-1G
APOSOR451190-1GEA 684 EUR
APOSOR451190-1G
(R)-1-[(Pyrrolidin-2-yl)methyl]piperidine dihydrochloride
(R)-1-[(Pyrrolidin-2-yl)methyl]piperidine dihydrochloride
Formel: C₁₀H₂₀N₂·2ClH
Molecular Weight: 241,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28134087
CAS-Nummer: 575469-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR