(R)-2-(Phenoxymethyl)-pyrrolidine hydrochloride

Lieferant: Apollo Scientific

OR451191-1G
APOSOR451191-1GEA 1160 EUR
APOSOR451191-1G
(R)-2-(Phenoxymethyl)-pyrrolidine hydrochloride
(R)-2-(Phenoxymethyl)-pyrrolidine hydrochloride
Formel: C₁₁H₁₅NO·ClH
Molecular Weight: 213,704 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13248613
CAS-Nummer: 1313254-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR