3-(Cyclopentyloxy)benzaldehyde

Lieferant: Apollo Scientific

OR451192-5G
APOSOR451192-5GEA 1190 EUR
APOSOR451192-5G
3-(Cyclopentyloxy)benzaldehyde
3-(Cyclopentyloxy)benzaldehyde
Formel: C₁₂H₁₄O₂
Molecular Weight: 190,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04037397
CAS-Nummer: 273722-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR