4-Cyclopentyl-benzenemethanol
Lieferant: Apollo Scientific
OR451194-1G
APOSOR451194-1GEA
1150
EUR
APOSOR451194-1G
4-Cyclopentyl-benzenemethanol
4-Cyclopentyl-benzenemethanol
Formel:
C₁₂H₁₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD17171062 CAS-Nummer: 171623-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...