4-(Cyclopentyloxy)-benzoic acid

Lieferant: Apollo Scientific

OR451195-5G
APOSOR451195-5GEA 960 EUR
APOSOR451195-5G
4-(Cyclopentyloxy)-benzoic acid
4-(Cyclopentyloxy)-benzoic acid
Formel: C₁₂H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03233617
CAS-Nummer: 30762-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR