Fmoc-2-cyclopentyl-glycine

Lieferant: Apollo Scientific

OR451196-1G
APOSOR451196-1GEA 771 EUR
APOSOR451196-1G
Fmoc-2-cyclopentyl-glycine
Fmoc-2-cyclopentyl-glycine
Formel: C₂₂H₂₃NO₄
Molecular Weight: 365,422 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08459479
CAS-Nummer: 1276562-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR