Ethyl 3-cyclopentyl-3-oxopropionate

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-cyclopentyl-3-oxopropanoate

OR451200-25G
APOSOR451200-25GEA 688 EUR
APOSOR451200-25G
Ethyl 3-cyclopentyl-3-oxopropionate
Ethyl 3-cyclopentyl-3-oxopropionate
Formel: C₁₀H₁₆O₃
Molecular Weight: 184,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410650
CAS-Nummer: 24922-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR