Boc-(R)-3-Cyclopentylalanine
Lieferant: Apollo Scientific
OR451201-1G
APOSOR451201-1GEA
0
EUR
APOSOR451201-1G
Boc-(R)-3-Cyclopentylalanine
Boc-(R)-3-Cyclopentylalanine
Formel:
C₁₃H₂₃NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094882 CAS-Nummer: 219819-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...