Boc-(S)-3-Cyclopentylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-((tert-Butoxycarbonyl)amino)-3-cyclopentylpropanoic acid

OR451202-1G
APOSOR451202-1GEA 140 EUR
APOSOR451202-1G
Boc-(S)-3-Cyclopentylalanine
Boc-(S)-3-Cyclopentylalanine
Formel: C₁₃H₂₃NO₄
Molecular Weight: 257,326 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094885
CAS-Nummer: 143415-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR