(S)-2-Amino-3-cyclopentylpropanoic acid 97%

Lieferant: Apollo Scientific

Synonyme: H-Cpa-OH, 3-Cyclopentyl-L-alanine

OR451203-1G
APOSOR451203-1GEA 202 EUR
APOSOR451203-1G
(S)-2-Amino-3-cyclopentylpropanoic acid 97%
(S)-2-Amino-3-cyclopentylpropanoic acid
Formel: C₈H₁₅NO₂
Molecular Weight: 157,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632069
CAS-Nummer: 99295-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR