3-Cyclopentane-D-Alanine

Lieferant: Apollo Scientific

Synonyme: H-D-3-Cyclopentane-Ala-OH, (R)-2-Amino-3-cyclopentylpropanoic acid

OR451204-1G
APOSOR451204-1GEA 730 EUR
APOSOR451204-1G
3-Cyclopentane-D-Alanine
3-Cyclopentane-D-Alanine
Formel: C₈H₁₅NO₂
Molecular Weight: 157.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788182
CAS-Nummer: 99295-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR