4-(Cyclopentyloxy)-benzeneacetic acid

Lieferant: Apollo Scientific

OR451208-1G
APOSOR451208-1GEA 771 EUR
APOSOR451208-1G
4-(Cyclopentyloxy)-benzeneacetic acid
4-(Cyclopentyloxy)-benzeneacetic acid
Formel: C₁₃H₁₆O₃
Molecular Weight: 220,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11131693
CAS-Nummer: 3585-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR