Boc-(R)-2-Cyclopentylglycine

Lieferant: Apollo Scientific

OR451209-1G
APOSOR451209-1GEA 0 EUR
APOSOR451209-1G
Boc-(R)-2-Cyclopentylglycine
Boc-(R)-2-Cyclopentylglycine
Formel: C₁₂H₂₁NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00671402
CAS-Nummer: 156881-63-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR