2-Cyano-N-cyclopentyl-acetamide
Lieferant: Apollo Scientific
OR451210-5G
APOSOR451210-5GEA
873
EUR
APOSOR451210-5G
2-Cyano-N-cyclopentyl-acetamide
2-Cyano-N-cyclopentyl-acetamide
Formel:
C₈H₁₂N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD01359391 CAS-Nummer: 15112-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...