2-Cyclopentylglycine
Lieferant: Apollo Scientific
Synonyme:
2-Amino-2-cyclopentylacetic acid
OR451212-5G
APOSOR451212-5GEA
161
EUR
APOSOR451212-5G
2-Cyclopentylglycine
2-Cyclopentylglycine
Formel:
C₇H₁₃NO₂ Molecular Weight: 143,184 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09264351 CAS-Nummer: 933-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...