3-Cyclopentyl-DL-alanine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-DL-Cpa-OH, H-β-Cyclopentyl-DL-Ala-OH

OR451213-1G
APOSOR451213-1GEA 300 EUR
APOSOR451213-1G
3-Cyclopentyl-DL-alanine
3-Cyclopentyl-DL-alanine
Formel: C₈H₁₅NO₂
Molecular Weight: 157,213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00237834
CAS-Nummer: 96539-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR