tert-Butyl-4-hydroxyazepane-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-hydroxyazepane-1-carboxylate, 1-Boc-4-Hydroxyazepane

OR451215-5G
APOSOR451215-5GEA 140 EUR
APOSOR451215-5G
tert-Butyl-4-hydroxyazepane-1-carboxylate
tert-Butyl-4-hydroxyazepane-1-carboxylate
Formel: C₁₁H₂₁NO₃
Molecular Weight: 215.2900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368610
CAS-Nummer: 478832-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR