1-Boc-2-[methoxy(methyl)carbamoyl]pyrrolidine
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 2-(methoxy(methyl)carbamoyl)pyrrolidine-1-carboxylate
OR451219-1G
APOSOR451219-1GEA
89.9
EUR
APOSOR451219-1G
1-Boc-2-[methoxy(methyl)carbamoyl]pyrrolidine
1-Boc-2-[methoxy(methyl)carbamoyl]pyrrolidine
Formel:
C₁₂H₂₂N₂O₄ Molecular Weight: 258.314 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04108777 CAS-Nummer: 334872-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...